Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTN2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266531.1 | NP_001253460.1 | 1020 | 123 | ||
XM_015135627.2 | XP_014991113.2 | 1049 | 123 | |||
XM_015135628.2 | XP_014991114.2 | 1081 | 123 | |||
XM_015135629.2 | XP_014991115.2 | 1154 | 123 | |||
XM_015135632.2 | XP_014991118.2 | 1150 | 123 | |||
XM_015135633.2 | XP_014991119.2 | 1150 | 123 | |||
GenBank mRNA | JU473960.1 | - | 1020 | 123 | ||
JU473961.1 | - | 1020 | 123 | |||
JU473963.1 | - | 1020 | 123 | |||
JU473964.1 | - | 1020 | 123 | |||
JU473966.1 | - | 1020 | 123 | |||
JU473967.1 | - | 1020 | 123 | |||
JU473968.1 | - | 1020 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |