Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLSCR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266547.1 | NP_001253476.1 | 945 | 72 | ||
GenBank mRNA | JU335565.1 | - | 977 | 72 | ||
JV045033.1 | - | 945 | 72 | |||
JV656521.1 | - | 975 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |