Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXXC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266606.1 | NP_001253535.1 | 1222 | 95 | ||
XM_015140813.2 | XP_014996299.1 | 1187 | 95 | |||
XM_015140814.2 | XP_014996300.1 | 1177 | 95 | |||
XM_015140816.2 | XP_014996302.1 | 1392 | 95 | |||
XM_015140818.2 | XP_014996304.1 | 1261 | 95 | |||
XM_028849040.1 | XP_028704873.1 | 2129 | 95 | |||
XM_028849041.1 | XP_028704874.1 | 1903 | 95 | |||
GenBank mRNA | JU334910.1 | - | 1220 | 95 | ||
JU475919.1 | - | 1215 | 95 | |||
JV047318.1 | - | 1222 | 95 | |||
JV686428.1 | - | 1223 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |