Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIM1K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266646.2 | NP_001253575.1 | 998 | 70 | ||
XM_015139962.2 | XP_014995448.1 | 1465 | 70 | |||
XM_015139963.2 | XP_014995449.1 | 1013 | 70 | |||
XM_028849135.1 | XP_028704968.1 | 1739 | 70 | |||
GenBank mRNA | JU333208.1 | - | 1001 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |