Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266674.1 | NP_001253603.1 | 1072 | 81 | ||
XM_015150824.2 | XP_015006310.1 | 1005 | 81 | |||
XM_028828993.1 | XP_028684826.1 | 1580 | 81 | |||
GenBank mRNA | JV667309.1 | - | 1072 | 81 | ||
JV669615.1 | - | 1072 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |