Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266679.1 | NP_001253608.1 | 395 | 65 | ||
GenBank mRNA | JU336175.1 | - | 488 | 65 | ||
JU477032.1 | - | 550 | 65 | |||
JV048126.1 | - | 395 | 65 | |||
JV048127.1 | - | 395 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |