Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEANC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266853.1 | NP_001253782.1 | 290 | 90 | ||
GenBank mRNA | JU332910.1 | - | 127 | 90 | ||
JU474306.1 | - | 290 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |