Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266867.1 | NP_001253796.1 | 226 | 75 | ||
GenBank mRNA | JU335229.1 | - | 220 | 75 | ||
JU476194.1 | - | 220 | 75 | |||
JV047501.1 | - | 226 | 75 | |||
JV602224.1 | - | 243 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |