Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIPK1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266885.1 | NP_001253814.1 | 363 | 113 | ||
GenBank mRNA | JU334451.1 | - | 311 | 113 | ||
JU475542.1 | - | 363 | 113 | |||
JV468120.1 | - | 340 | 113 | |||
JV616274.1 | - | 369 | 113 | |||
JV683580.1 | - | 345 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |