Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266918.1 | NP_001253847.1 | 1819 | 65 | ||
GenBank mRNA | JU327277.1 | - | 1457 | 65 | ||
JU472720.1 | - | 1666 | 65 | |||
JV043808.1 | - | 1819 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |