Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289897.1 | NP_001276826.1 | 704 | 62 | ||
GenBank mRNA | JV708635.1 | - | 704 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Macaca mulatta |