Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278446.1 | NP_001265375.1 | 404 | 56 | ||
GenBank mRNA | AY950561.1 | - | 88 | 56 | ||
JU335774.1 | - | 348 | 56 | |||
JU473363.1 | - | 345 | 56 | |||
JV515827.1 | - | 404 | 56 | |||
JV589729.1 | - | 391 | 56 | |||
JV720426.1 | - | 348 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |