Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260572.2 | NP_001247501.1 | 898 | 76 | ||
XM_028846130.1 | XP_028701963.1 | 769 | 76 | |||
GenBank mRNA | JU337265.1 | - | 908 | 76 | ||
JU477971.1 | - | 898 | 76 | |||
JV528725.1 | - | 875 | 76 | |||
JV599297.1 | - | 909 | 76 | |||
JV634423.1 | - | 882 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |