Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761975.1 | XP_008760197.1 | 6 - 7 | 2291 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |