Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012827.2 | NP_036959.2 | 506 | 61 | ||
XM_006251746.2 | XP_006251808.1 | 1268 | 61 | |||
XM_006251747.2 | XP_006251809.1 | 818 | 61 | |||
XM_006251748.2 | XP_006251810.1 | 3 - 4 | 800 | 61 | ||
XM_006251749.2 | XP_006251811.1 | 542 | 61 | |||
XM_006251751.2 | XP_006251813.1 | 519 | 61 | |||
XM_008770501.1 | XP_008768723.1 | 632 | 61 | |||
GenBank mRNA | AY184241.1 | - | 1 - 2 | 368 | 61 | |
BC078901.1 | - | 506 | 7121295 | 61 | ||
Z22607.1 | - | 3 - 4 | 768 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |