Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107409.1 | NP_001100879.1 | 2 - 3 | 216 | 108 | |
XM_006255082.1 | XP_006255144.1 | 2 - 3 | 216 | 108 | ||
GenBank mRNA | BC168200.1 | - | 2 - 3 | 232 | 7114331 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |