Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107895.2 | NP_001101365.2 | 3 - 4 | 287 | 81 | |
GenBank mRNA | CN544042.1 | - | 1 - 2 | 81 | ||
FM087109.1 | - | 3 - 4 | 287 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |