Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107237.1 | NP_001100707.1 | 3 - 4 | 369 | 63 | |
XM_008770315.1 | XP_008768537.1 | 3 - 4 | 709 | 63 | ||
XM_008770317.1 | XP_008768539.1 | 3 - 4 | 709 | 63 | ||
XM_008770318.1 | XP_008768540.1 | 3 - 4 | 709 | 63 | ||
XM_008770319.1 | XP_008768541.1 | 3 - 4 | 709 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Rattus norvegicus Probe spans exons |