Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177931.2 | NP_808792.1 | 14 - 15 | 2262 | 97 | |
XM_006238515.2 | XP_006238577.1 | 15 - 16 | 2464 | 97 | ||
XM_006238516.2 | XP_006238578.1 | 16 - 17 | 2577 | 97 | ||
GenBank mRNA | AB105378.1 | - | 14 - 15 | 2262 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |