Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snap25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030991.3 | NP_112253.1 | 372 | 71 | ||
GenBank mRNA | AB003992.1 | - | 163 | 71 | ||
AF245227.1 | - | 371 | 71 | |||
BC087699.1 | - | 327 | 7314318 | 71 | ||
FQ212040.1 | - | 343 | 71 | |||
FQ213289.1 | - | 4 | 340 | 71 | ||
FQ213923.1 | - | 343 | 71 | |||
FQ214162.1 | - | 372 | 71 | |||
U56262.1 | - | 136 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |