Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199376.1 | NP_955408.1 | 1 - 2 | 68 | 85 | |
GenBank mRNA | BC062050.1 | - | 1 - 2 | 68 | 5598825 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |