Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108285.1 | NP_001101755.1 | 3 - 4 | 490 | 90 | |
XM_006246907.1 | XP_006246969.1 | 3 - 4 | 690 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |