Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191683.1 | NP_001178612.1 | 2 - 3 | 629 | 152 | |
XM_006251579.2 | XP_006251641.2 | 2 - 3 | 415 | 152 | ||
XM_006251587.2 | XP_006251649.1 | 2 - 3 | 550 | 152 | ||
XM_006251588.2 | XP_006251650.1 | 2 - 3 | 851 | 152 | ||
XM_006251594.2 | XP_006251656.1 | 2 - 3 | 838 | 152 | ||
XM_008770430.1 | XP_008768652.1 | 2 - 3 | 851 | 152 | ||
GenBank mRNA | BC086607.1 | - | 2 - 3 | 738 | 152 | |
BC158689.1 | - | 2 - 3 | 542 | 8375842 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |