Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024140.2 | NP_077054.1 | 1 - 2 | 263 | 63 | |
GenBank mRNA | FQ213074.1 | - | 1 - 2 | 61 | 63 | |
L09119.1 | - | 1 - 2 | 263 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus 1700 Inventoried Probe spans exons |