Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100840.1 | NP_001094310.1 | 3 - 4 | 350 | 135 | |
GenBank mRNA | BC100255.1 | - | 3 - 4 | 292 | 135 | |
BC158737.1 | - | 3 - 4 | 296 | 135 | ||
CV117277.1 | - | 3 - 4 | 587 | 135 | ||
FQ218354.1 | - | 3 - 4 | 247 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |