Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013066.1 | NP_037198.1 | 6 - 7 | 4496 | 79 | |
XM_006245100.2 | XP_006245162.1 | 6 - 7 | 8645 | 79 | ||
XM_008767197.1 | XP_008765419.1 | 6 - 7 | 4676 | 79 | ||
XM_008767198.1 | XP_008765420.1 | 6 - 7 | 4676 | 79 | ||
XM_008767199.1 | XP_008765421.1 | 6 - 7 | 4673 | 79 | ||
XM_008767201.1 | XP_008765423.1 | 6 - 7 | 4673 | 79 | ||
XM_008767202.1 | XP_008765424.1 | 6 - 7 | 4664 | 79 | ||
XM_008767203.1 | XP_008765425.1 | 6 - 7 | 4676 | 79 | ||
XM_008767204.1 | XP_008765426.1 | 5 - 6 | 4430 | 79 | ||
XM_008767205.1 | XP_008765427.1 | 5 - 6 | 4418 | 79 | ||
XM_008767206.1 | XP_008765428.1 | 6 - 7 | 4464 | 79 | ||
GenBank mRNA | X54100.1 | - | 6 - 7 | 4496 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |