Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053662.3 | NP_446114.1 | 697 | 89 | ||
XM_008761029.1 | XP_008759251.1 | 1773 | 89 | |||
GenBank mRNA | AF030091.3 | - | 697 | 89 | ||
BC085686.1 | - | 751 | 89 | |||
BC128699.1 | - | 791 | 89 | |||
BC161817.1 | - | 643 | 9022416 | 89 | ||
FQ213168.1 | - | 1 - 2 | 121 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |