Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008319.2 | NP_001008320.2 | 2 - 3 | 289 | 73 | |
GenBank mRNA | BC085770.1 | - | 2 - 3 | 260 | 7189726 | 73 |
CV109439.1 | - | 2 - 3 | 289 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |