Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Midn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191577.1 | NP_001178506.1 | 1 - 2 | 238 | 64 | |
XM_006240911.2 | XP_006240973.1 | 2 - 3 | 1895 | 64 | ||
XM_006240912.2 | XP_006240974.1 | 2 - 3 | 1622 | 64 | ||
XM_006240913.2 | XP_006240975.1 | 2 - 3 | 1895 | 64 | ||
XM_006240914.2 | XP_006240976.1 | 2 - 3 | 1895 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |