Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109292.1 | NP_001102762.1 | 11 - 12 | 1524 | 105 | |
XM_006242950.2 | XP_006243012.1 | 11 - 12 | 1524 | 105 | ||
XM_006242951.2 | XP_006243013.1 | 11 - 12 | 1538 | 105 | ||
XM_006242952.2 | XP_006243014.1 | 11 - 12 | 1533 | 105 | ||
XM_006242954.2 | XP_006243016.1 | 11 - 12 | 1807 | 105 | ||
GenBank mRNA | FQ234306.1 | - | 3 - 4 | 319 | 105 | |
FQ235180.1 | - | 3 - 4 | 309 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |