Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191070.1 | NP_001177999.1 | 3 - 4 | 966 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons |