Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wiz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108064.1 | NP_001101534.1 | 2 - 3 | 305 | 62 | |
XM_008765167.1 | XP_008763389.1 | 2 - 3 | 768 | 62 | ||
XM_008765168.1 | XP_008763390.1 | 2 - 3 | 768 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |