Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il36rn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107814.1 | NP_001101284.1 | 3 - 4 | 348 | 89 | |
XM_008761586.1 | XP_008759808.1 | 1 - 2 | 999 | 89 | ||
XM_008761587.1 | XP_008759809.1 | 2 - 3 | 642 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |