Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qki | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008758777.1 | XP_008756999.1 | 4 - 5 | 1004 | 67 | |
XM_008758779.1 | XP_008757001.1 | 1004 | 67 | |||
XM_008758780.1 | XP_008757002.1 | 1003 | 67 |
Gene Symbol: Qk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115021.1 | NP_001108493.1 | 6 - 7 | 864 | 67 | |
GenBank mRNA | AB054997.1 | - | 3 - 4 | 342 | 67 | |
BC158800.1 | - | 6 - 7 | 864 | 7320367 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |