Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014159.2 | NP_001014181.2 | 4 - 5 | 528 | 68 | |
GenBank mRNA | BC079321.1 | - | 4 - 5 | 486 | 7135399 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |