Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106061.1 | NP_001099531.1 | 8 - 9 | 1163 | 71 | |
XM_006252574.2 | XP_006252636.1 | 11 - 12 | 1295 | 71 | ||
XM_006252575.2 | XP_006252637.1 | 11 - 12 | 1493 | 71 | ||
XM_006252576.2 | XP_006252638.1 | 9 - 10 | 1659 | 71 | ||
XM_008770982.1 | XP_008769204.1 | 11 - 12 | 1360 | 71 | ||
XM_008770983.1 | XP_008769205.1 | 11 - 12 | 1202 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |