Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106165.1 | NP_001099635.1 | 1 - 2 | 134 | 66 | |
XM_008772243.1 | XP_008770465.1 | 1 - 2 | 96 | 66 | ||
GenBank mRNA | BC167038.1 | - | 1 - 2 | 103 | 9090629 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |