Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012492.2 | NP_036624.2 | 1 | 649 | 61 | |
GenBank mRNA | BC086538.1 | - | 1 | 649 | 7114502 | 61 |
J03024.1 | - | 1 | 551 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Both primers and probe map within a single exon |