Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012523.2 | NP_036655.1 | 3 - 4 | 399 | 72 | |
GenBank mRNA | BC078900.1 | - | 3 - 4 | 350 | 7130719 | 72 |
M57276.1 | - | 3 - 4 | 413 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |