Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012529.3 | NP_036661.3 | 1051 | 82 | ||
GenBank mRNA | BC065307.1 | - | 1033 | 5598839 | 82 | |
BC070955.1 | - | 1038 | 7104205 | 82 | ||
BC087656.1 | - | 1050 | 82 | |||
BC099814.1 | - | 1035 | 7929525 | 82 | ||
BC127477.1 | - | 1041 | 8367180 | 82 | ||
M14400.1 | - | 1063 | 82 | |||
M57664.1 | - | 6 - 7 | 914 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |