Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Comt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012531.2 | NP_036663.1 | 4 - 5 | 757 | 75 | |
XM_006248603.2 | XP_006248665.1 | 4 - 5 | 758 | 75 | ||
GenBank mRNA | BC081850.1 | - | 4 - 5 | 615 | 7191956 | 75 |
FQ209401.1 | - | 3 - 4 | 470 | 75 | ||
FQ209488.1 | - | 3 - 4 | 476 | 75 | ||
FQ209706.1 | - | 3 - 4 | 481 | 75 | ||
FQ210416.1 | - | 3 - 4 | 498 | 75 | ||
FQ210430.1 | - | 3 - 4 | 473 | 75 | ||
FQ218190.1 | - | 3 - 4 | 472 | 75 | ||
FQ218375.1 | - | 3 - 4 | 480 | 75 | ||
FQ218564.1 | - | 3 - 4 | 472 | 75 | ||
FQ218595.1 | - | 3 - 4 | 472 | 75 | ||
FQ219155.1 | - | 3 - 4 | 477 | 75 | ||
FQ219257.1 | - | 3 - 4 | 477 | 75 | ||
FQ219433.1 | - | 3 - 4 | 472 | 75 | ||
FQ219709.1 | - | 1 - 2 | 123 | 75 | ||
FQ223536.1 | - | 3 - 4 | 472 | 75 | ||
FQ228829.1 | - | 3 - 4 | 470 | 75 | ||
FQ228870.1 | - | 3 - 4 | 478 | 75 | ||
FQ229523.1 | - | 3 - 4 | 452 | 75 | ||
M60753.1 | - | 561 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |