Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012589.2 | NP_036721.1 | 2 - 3 | 270 | 128 | |
GenBank mRNA | M26744.1 | - | 2 - 3 | 270 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Rattus norvegicus Rat Androgen Panel Rat Chemokine Panel Inventoried Probe spans exons |