Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ucp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012682.2 | NP_036814.1 | 2 - 3 | 502 | 69 | |
GenBank mRNA | BC088156.1 | - | 2 - 3 | 490 | 7380759 | 69 |
FM087156.1 | - | 2 - 3 | 501 | 69 | ||
M11814.1 | - | 2 - 3 | 500 | 69 | ||
X03894.1 | - | 2 - 3 | 412 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |