Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amacr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012816.2 | NP_036948.2 | 3 - 4 | 562 | 124 | |
GenBank mRNA | BC078885.1 | - | 3 - 4 | 530 | 7110324 | 124 |
BC091163.1 | - | 3 - 4 | 531 | 7130642 | 124 | |
BC127505.1 | - | 3 - 4 | 551 | 7325347 | 124 | |
U89905.1 | - | 3 - 4 | 550 | 124 | ||
Y08172.1 | - | 3 - 4 | 540 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |