Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfya | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012865.1 | NP_036997.1 | 3 - 4 | 339 | 72 | |
XM_006244409.2 | XP_006244471.1 | 3 - 4 | 346 | 72 | ||
XM_006244410.1 | XP_006244472.1 | 3 - 4 | 342 | 72 | ||
XM_008766839.1 | XP_008765061.1 | 2 - 3 | 242 | 72 | ||
GenBank mRNA | FQ223023.1 | - | 3 - 4 | 351 | 72 | |
M34238.1 | - | 3 - 4 | 339 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus 1700 Inventoried Probe spans exons |