Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Id2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013060.3 | NP_037192.1 | 1 - 2 | 409 | 70 | |
GenBank mRNA | BC086391.1 | - | 404 | 7303450 | 70 | |
D10863.1 | - | 1 - 2 | 409 | 70 | ||
FQ212969.1 | - | 427 | 70 | |||
FQ213471.1 | - | 427 | 70 | |||
FQ213662.1 | - | 427 | 70 | |||
FQ220574.1 | - | 1 - 2 | 427 | 70 | ||
FQ227397.1 | - | 1 - 2 | 427 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |