Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fabp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017098.1 | NP_058794.1 | 1 - 2 | 111 | 98 | |
GenBank mRNA | L22788.1 | - | 1 - 2 | 111 | 98 | |
S52878.1 | - | 1 - 2 | 90 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus 1700 Inventoried Probe spans exons |