Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017240.2 | NP_058936.1 | 37 - 38 | 5798 | 66 | |
XM_006251951.2 | XP_006252013.1 | 39 - 40 | 5948 | 66 | ||
GenBank mRNA | FQ215585.1 | - | 6 - 7 | 1062 | 66 | |
FQ216901.1 | - | 6 - 7 | 938 | 66 | ||
FQ216969.1 | - | 3 - 4 | 311 | 66 | ||
FQ217025.1 | - | 4 - 5 | 599 | 66 | ||
FQ217049.1 | - | 3 - 4 | 515 | 66 | ||
FQ217078.1 | - | 4 - 5 | 602 | 66 | ||
FQ217204.1 | - | 1 - 2 | 83 | 66 | ||
FQ217214.1 | - | 8 - 9 | 1452 | 66 | ||
FQ217258.1 | - | 3 - 4 | 431 | 66 | ||
FQ217267.1 | - | 4 - 5 | 599 | 66 | ||
FQ217344.1 | - | 4 - 5 | 602 | 66 | ||
FQ217392.1 | - | 6 - 7 | 1050 | 66 | ||
FQ217405.1 | - | 4 - 5 | 599 | 66 | ||
FQ217522.1 | - | 6 - 7 | 935 | 66 | ||
FQ217648.1 | - | 1 - 2 | 29 | 66 | ||
FQ217657.1 | - | 2 - 3 | 176 | 66 | ||
FQ217776.1 | - | 1 - 2 | 80 | 66 | ||
FQ217816.1 | - | 4 - 5 | 602 | 66 | ||
FQ217848.1 | - | 4 - 5 | 602 | 66 | ||
FQ217941.1 | - | 3 - 4 | 356 | 66 | ||
FQ217965.1 | - | 4 - 5 | 599 | 66 | ||
FQ218011.1 | - | 4 - 5 | 536 | 66 | ||
FQ218024.1 | - | 3 - 4 | 311 | 66 | ||
FQ223551.1 | - | 3 - 4 | 431 | 66 | ||
FQ223587.1 | - | 3 - 4 | 356 | 66 | ||
FQ223806.1 | - | 4 - 5 | 599 | 66 | ||
FQ223821.1 | - | 4 - 5 | 602 | 66 | ||
FQ223892.1 | - | 4 - 5 | 599 | 66 | ||
FQ223928.1 | - | 3 - 4 | 282 | 66 | ||
FQ223936.1 | - | 4 - 5 | 532 | 66 | ||
FQ223945.1 | - | 7 - 8 | 1175 | 66 | ||
FQ224001.1 | - | 3 - 4 | 359 | 66 | ||
FQ224034.1 | - | 2 - 3 | 176 | 66 | ||
FQ224069.1 | - | 3 - 4 | 311 | 66 | ||
FQ224090.1 | - | 3 - 4 | 431 | 66 | ||
FQ224098.1 | - | 3 - 4 | 431 | 66 | ||
FQ224119.1 | - | 4 - 5 | 527 | 66 | ||
FQ224260.1 | - | 5 - 6 | 814 | 66 | ||
FQ224368.1 | - | 3 - 4 | 356 | 66 | ||
FQ224375.1 | - | 2 - 3 | 223 | 66 | ||
FQ224416.1 | - | 3 - 4 | 431 | 66 | ||
FQ224696.1 | - | 4 - 5 | 599 | 66 | ||
FQ224790.1 | - | 3 - 4 | 260 | 66 | ||
X15939.1 | - | 37 - 38 | 5800 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |