Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017250.1 | NP_058946.1 | 2 - 3 | 780 | 82 | |
XM_008767216.1 | XP_008765438.1 | 3 - 4 | 1630 | 82 | ||
GenBank mRNA | X66842.1 | - | 2 - 3 | 780 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons |