Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsk3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017344.1 | NP_059040.1 | 4 - 5 | 794 | 69 | |
XM_008758972.1 | XP_008757194.1 | 4 - 5 | 803 | 69 | ||
GenBank mRNA | X53427.1 | - | 4 - 5 | 794 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Rattus norvegicus Probe spans exons |